Learn More
Medchemexpress LLC Lucifer Yellow CH dipotassium salt | 71206-95-6 | MFCD00013445 | 99.8% | 521.57 | C13H9K2N5O9S2 | 10 MG
SDP
Catalog No. 5000283609
Encompass_Preferred
Lucifer Yellow CH dipotassium salt is a high-intensity fluorescent tracer used in neuroscience and cell biology for intracellular labeling, neuronal tracing, gap junction analysis, and selective cell visualization. Supplied as the dipotassium salt with high reported purity, it is suitable for fluorescence microscopy and experiments requiring reliable, fixable fluorescent markers.
  • High-intensity fluorescence for sensitive detection.
  • Useful for neuronal tracing and monitoring neuronal branching.
  • Enables gap junction detection and characterization.
  • Reactive hydrazyl groups allow fixation and labeling with aldehyde-based methods.
  • Suitable for fluorescence microscopy and electrophysiological assays.
Catalog No. 50-002-83609 Supplier Medchemexpress LLC Supplier No. HYW12771510MG
Add to Cart
Add to Cart
Lucifer Yellow CH dipotassium salt is a high-intensity fluorescent tracer used in neuroscience and cell biology for intracellular labeling, neuronal tracing, gap junction analysis, and selective cell visualization. Supplied as the dipotassium salt with high reported purity, it is suitable for fluorescence microscopy and experiments requiring reliable, fixable fluorescent markers.
  • High-intensity fluorescence for sensitive detection.
  • Useful for neuronal tracing and monitoring neuronal branching.
  • Enables gap junction detection and characterization.
  • Reactive hydrazyl groups allow fixation and labeling with aldehyde-based methods.
  • Suitable for fluorescence microscopy and electrophysiological assays.