Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lucifer yellow CH dipotassium salt | 71206-95-6 | MFCD00013445 | 99.8% | 521.57 g/mol | C13H9K2N5O9S2 | 1 ML
SDP

Catalog No. 5000282691
Encompass_Preferred

Lucifer Yellow CH dipotassium salt is a high-intensity fluorescent tracer used for neuronal labeling, gap junction detection, and selective cell ablation after aldehyde fixation. Supplied as a solid or as a 10 mM solution in DMSO, it provides bright fluorescence for microscopy and cell tracing applications.

  • High-intensity fluorescent probe for neuronal tracing and gap junction studies.
  • Excitation approximately 430-450 nm and emission approximately 540-650 nm.
  • Available as 10 mM solution (1 mL) in DMSO or multiple solid sizes.
  • High purity (~99.8%) suitable for analytical and biological experiments.
  • Storage recommendations: 4°C sealed; in solvent -80°C for long-term stability.

Catalog No. 50-002-82691 Supplier Medchemexpress LLC Supplier No. HYW12771510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lucifer Yellow CH dipotassium salt is a high-intensity fluorescent tracer used for neuronal labeling, gap junction detection, and selective cell ablation after aldehyde fixation. Supplied as a solid or as a 10 mM solution in DMSO, it provides bright fluorescence for microscopy and cell tracing applications.

  • High-intensity fluorescent probe for neuronal tracing and gap junction studies.
  • Excitation approximately 430-450 nm and emission approximately 540-650 nm.
  • Available as 10 mM solution (1 mL) in DMSO or multiple solid sizes.
  • High purity (~99.8%) suitable for analytical and biological experiments.
  • Storage recommendations: 4°C sealed; in solvent -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.