Learn More
Biotium Lucifer Yellow CH Potassium Salt
Catalog No. 501964783
Encompass_Preferred
Lucifer Yellow CH potassium salt (LY CH potassium salt) is a widely used polar molecular tracer for studying neuronal morphology. The fluorescent molecule contains a carbohydrazide that allow the molecule to be aldehyde-fixable. Properties: Ex/ Em = 428/536 nm; Yellow solid soluble in water; Store at 4°C and protect from light; C13H9K2N5O9S2; MW: 522; [71206-95-6]
Catalog No. 50-196-4783 Supplier Biotium Supplier No. 80016
Lucifer Yellow CH potassium salt (LY CH potassium salt) is a widely used polar molecular tracer for studying neuronal morphology. The fluorescent molecule contains a carbohydrazide that allow the molecule to be aldehyde-fixable. Properties: Ex/ Em = 428/536 nm; Yellow solid soluble in water; Store at 4°C and protect from light; C13H9K2N5O9S2; MW: 522; [71206-95-6]