Learn More
Cayman Chemical Lucifer Yellow CH, Purity: ≥80%, CAS: 67769-47-5, 10 mg
SDP
Catalog No. 503015920
Encompass_Preferred
A fluorescent probe; contains a free hydrazido group that reacts with aliphatic aldehydes at room temperature; ex/em = 430/540 nm, respectively; been used as a biological tracer to monitor the branching pattern and regeneration of neurons, gap junction detection and characterization, and selective ablation of cells following aldehyde fixation
Catalog No. 50-301-5920 Supplier Cayman Chemical Supplier No. 2557310 MG
A fluorescent probe; contains a free hydrazido group that reacts with aliphatic aldehydes at room temperature; ex/em = 430/540 nm, respectively; been used as a biological tracer to monitor the branching pattern and regeneration of neurons, gap junction detection and characterization, and selective ablation of cells following aldehyde fixation