Learn More
Electron Microscopy Sciences Lucifer Yellow, CH, Dilithium Salt 25 MG
SDP
Catalog No. 5027006
Encompass_Preferred
CAS #67769-47-5 C₁₃H₉Li₂N₂O₉S₂ F.W. 457.25
A technique producing an electron dense osmiophilic polymer when Lucifer Yellow is injected into cells. Lucifer Yellow also can be viewed in living cells as a dye marker and is more sensitive than any other dye. See Cell Tissue Res., 204, 37 (1979) J. Histochem Cystochem, 30, 189 (1982) Science, 217, 953 (1982); 215, 1641 (1982); 214, 801 (1981); 225, 851 (1984); Cell, 14, 741 (1978) or A fluorescent dye. Nature 292, 17 (1981)
Catalog No. 50-270-06 Supplier Electron Microscopy Sciences Supplier No. 17950
CAS #67769-47-5 C₁₃H₉Li₂N₂O₉S₂ F.W. 457.25
A technique producing an electron dense osmiophilic polymer when Lucifer Yellow is injected into cells. Lucifer Yellow also can be viewed in living cells as a dye marker and is more sensitive than any other dye. See Cell Tissue Res., 204, 37 (1979) J. Histochem Cystochem, 30, 189 (1982) Science, 217, 953 (1982); 215, 1641 (1982); 214, 801 (1981); 225, 851 (1984); Cell, 14, 741 (1978) or A fluorescent dye. Nature 292, 17 (1981)