Learn More
Amsbio LLC Luciferase (EF1a Promoter, Bsd), Concentrated Lentivirus, Size 1x10^8 IFU/ml x 200ul
Catalog No. NC3913737
Encompass
Pre-made Lentivirus expressing firefly luciferase under EF1a promoter with Blastidicin selection marker, provided in PBS solution.
Catalog No. NC3913737 Supplier Amsbio LLC Supplier No. LVP433PBS
Pre-made Lentivirus expressing firefly luciferase under EF1a promoter with Blastidicin selection marker, provided in PBS solution.