Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Luciferase-in-1 | 10205-56-8 | 99.6% | C15H14N2S | 10MG
SDP

Catalog No. 5000259174
Encompass_Preferred

Luciferase-IN-1 is a potent small-molecule inhibitor of firefly luciferase used as a biochemical tool in in vitro and in vivo assay systems. It is supplied as a characterized solid with recommended storage and formulation guidance for preparing stock solutions.

  • Potent luciferase inhibitor for biochemical assays.
  • High purity (99.6%) suitable for research applications.
  • Supplied as a solid, light yellow to brown in appearance.
  • Soluble in DMSO; stock solutions commonly prepared at 10 mg/mL.
  • Storage recommendations provided for powder and solutions.
  • Includes in vivo formulation examples for administration.
  • Intended for laboratory research use only, not for human use.

Catalog No. 50-002-59174 Supplier Medchemexpress LLC Supplier No. HY13670610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luciferase-IN-1 is a potent small-molecule inhibitor of firefly luciferase used as a biochemical tool in in vitro and in vivo assay systems. It is supplied as a characterized solid with recommended storage and formulation guidance for preparing stock solutions.

  • Potent luciferase inhibitor for biochemical assays.
  • High purity (99.6%) suitable for research applications.
  • Supplied as a solid, light yellow to brown in appearance.
  • Soluble in DMSO; stock solutions commonly prepared at 10 mg/mL.
  • Storage recommendations provided for powder and solutions.
  • Includes in vivo formulation examples for administration.
  • Intended for laboratory research use only, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.