Learn More
Creative Biogene LUCIFERASE REP CL OVCAR8
SDP
Catalog No. NC3452834
Encompass
  • Meticulously engineered human cell line designed for advanced cancer research.
  • Stable overexpression of luciferase reporter gene.
  • Enables quantitative assessment of transcriptional activity.
  • Real-time monitoring of gene expression and signaling pathways.
  • Invaluable tool for studying cancer biology including tumor growth.
  • Appropriate for metastasis and drug response studies.
  • Ideal for high-throughput screening assays.
  • Enhanced sensitivity for detecting molecular events.
  • Validated stability for at least 10 passages.
  • Negative for bacteria, yeast, fungi, and mycoplasma.
  • Consistent reporter construct stably integrated into genome.
  • Induces strong reporter signal in response to stimuli.
  • Facilitates live cell imaging and drug screening.
  • Applicable to research on ovarian cancer.
Catalog No. NC3452834 Supplier Creative Biogene Supplier No. CSCRR0546
Add to Cart
Add to Cart
Request a Quote
  • Meticulously engineered human cell line designed for advanced cancer research.
  • Stable overexpression of luciferase reporter gene.
  • Enables quantitative assessment of transcriptional activity.
  • Real-time monitoring of gene expression and signaling pathways.
  • Invaluable tool for studying cancer biology including tumor growth.
  • Appropriate for metastasis and drug response studies.
  • Ideal for high-throughput screening assays.
  • Enhanced sensitivity for detecting molecular events.
  • Validated stability for at least 10 passages.
  • Negative for bacteria, yeast, fungi, and mycoplasma.
  • Consistent reporter construct stably integrated into genome.
  • Induces strong reporter signal in response to stimuli.
  • Facilitates live cell imaging and drug screening.
  • Applicable to research on ovarian cancer.