Learn More
AdipoGen Lucigenin
SDP
Catalog No. 502561266
Encompass_Preferred
Chemical. CAS 2315-97-1. Formula C28H22N4O6. MW 510.5. Synthetic. Chemiluminescent probe for the detection of peroxides in biological systems. Reagent used for the generation of chemiluminescence in the presence of a reductant and oxygen It is employed in numerous chemiluminescent assays. Lucigenin is specific for superoxide anion radical.
Catalog No. 50-256-1266 Supplier AdipoGen Supplier No. CDXD0068G002
Chemical. CAS 2315-97-1. Formula C28H22N4O6. MW 510.5. Synthetic. Chemiluminescent probe for the detection of peroxides in biological systems. Reagent used for the generation of chemiluminescence in the presence of a reductant and oxygen It is employed in numerous chemiluminescent assays. Lucigenin is specific for superoxide anion radical.