Learn More
Abcam Lucigenin, Supero x ide anion radical chemiluminescent probe, 10G
Catalog No. 502705300
Encompass_Preferred
MW 510.5 Da, Purity >98%. Selective superoxide anion radical chemiluminescent probe and fluorescent chloride-sensitive indicator. Generates chemiluminescence in the presence of superoxide (O₂-). Detects vascular superoxide formation in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5300 Supplier Abcam Supplier No. AB14531010G
MW 510.5 Da, Purity >98%. Selective superoxide anion radical chemiluminescent probe and fluorescent chloride-sensitive indicator. Generates chemiluminescence in the presence of superoxide (O₂-). Detects vascular superoxide formation in vivo..
The product is subject to the following: Abcam Restricted Use Statement