Learn More
Medchemexpress LLC Lucitanib | 1058137-23-7 | C26H25N3O4 | 100 MG
SDP
Catalog No. 5000376529
Encompass_Preferred
Lucitanib (E-3810) is a novel dual inhibitor of VEGFR and FGFR, potently and selectively inhibiting multiple receptors. It is suitable for use in various laboratory applications, including the manufacture of substances.
  • Inhibits VEGFR1 with an IC50 of 7 nM
  • Inhibits VEGFR2 with an IC50 of 25 nM
  • Inhibits VEGFR3 with an IC50 of 10 nM
  • Inhibits FGFR1 with an IC50 of 17.5 nM
  • Inhibits FGFR2 with an IC50 of 82.5 nM
  • Targeted for cancer research
Catalog No. 50-003-76529 Supplier Medchemexpress LLC Supplier No. HY15391100MG
Lucitanib (E-3810) is a novel dual inhibitor of VEGFR and FGFR, potently and selectively inhibiting multiple receptors. It is suitable for use in various laboratory applications, including the manufacture of substances.
  • Inhibits VEGFR1 with an IC50 of 7 nM
  • Inhibits VEGFR2 with an IC50 of 25 nM
  • Inhibits VEGFR3 with an IC50 of 10 nM
  • Inhibits FGFR1 with an IC50 of 17.5 nM
  • Inhibits FGFR2 with an IC50 of 82.5 nM
  • Targeted for cancer research