Learn More
Electron Microscopy Sciences EMS Lucite 12-Film Rack, for 4x5"
SDP
Catalog No. 5025790
Encompass_Preferred
Fabricated from genuine Lucite; these racks fit into our 5"x7" developing tanks (EMS Cat. ). One end of the rack is extended in length with a cut-out hole to serve as a handle. The racks are designed to accommodate a number of sheets per rack without changing the length of the rack (As the number of the films per rack increases, the space between them decreases).
Use: 4 x 5" films
Size: 19.2 x 10.4 x 20 cm
Catalog No. 50-257-90 Supplier Electron Microscopy Sciences Supplier No. 7489512
Fabricated from genuine Lucite; these racks fit into our 5"x7" developing tanks (EMS Cat. ). One end of the rack is extended in length with a cut-out hole to serve as a handle. The racks are designed to accommodate a number of sheets per rack without changing the length of the rack (As the number of the films per rack increases, the space between them decreases).
Use: 4 x 5" films
Size: 19.2 x 10.4 x 20 cm