Learn More
Electron Microscopy Sciences EMS Lucite Rack for 18 Plates, For 3-1/4 x4"
SDP
Catalog No. 5025788
Encompass_Preferred
Fabricated from genuine Lucite; these racks fit into our 5"x7" developing tanks (EMS Cat. ). One end of the rack is extended in length with a cut-out hole to serve as a handle. The racks are designed to accommodate a number of sheets per rack without changing the length of the rack (As the number of the films per rack increases, the space between them decreases).
Use: 3-1/4 x 4" films
Size: 19.2 cm x 8.5 cm x 20 cm
Catalog No. 50-257-88 Supplier Electron Microscopy Sciences Supplier No. 7489018
Fabricated from genuine Lucite; these racks fit into our 5"x7" developing tanks (EMS Cat. ). One end of the rack is extended in length with a cut-out hole to serve as a handle. The racks are designed to accommodate a number of sheets per rack without changing the length of the rack (As the number of the films per rack increases, the space between them decreases).
Use: 3-1/4 x 4" films
Size: 19.2 cm x 8.5 cm x 20 cm