Learn More
QA Bio Inc LudgerClean A cartridges
Catalog No. 502236696
Encompass
Hydrophilic interaction amide cartridges • For purification of 2AA or 2AB labelled N glycans, VTag labelled glycopeptides or APTS labelled N-glycans • Each pack contains 24 cartridges • Can be used in a vacuum manifold system (e.g. Ludger LV system) that can process up to 96 cartridges simultaneously.
This item is not returnable. View return policy
Catalog No. 50-223-6696 Supplier QA Bio Inc Supplier No. LCA24
Hydrophilic interaction amide cartridges • For purification of 2AA or 2AB labelled N glycans, VTag labelled glycopeptides or APTS labelled N-glycans • Each pack contains 24 cartridges • Can be used in a vacuum manifold system (e.g. Ludger LV system) that can process up to 96 cartridges simultaneously.