Learn More
QA Bio Inc LudgerClean T1 cartridges - 6 cartridges
Catalog No. 502236707
Encompass
Hydrophilic interaction silica cartridges for post-labeling purification of LudgerTag™ fluorophore and chromophore labeled glycans. • Suitable for cleanup after glycan labeling with 2-AB (2-aminobenzamide) and 2-AA (2-aminobenzoic acid). • Each pack contains 6 cartridges.
This item is not returnable. View return policy
Catalog No. 50-223-6707 Supplier QA Bio Inc Supplier No. LCT1A6
Hydrophilic interaction silica cartridges for post-labeling purification of LudgerTag™ fluorophore and chromophore labeled glycans. • Suitable for cleanup after glycan labeling with 2-AB (2-aminobenzamide) and 2-AA (2-aminobenzoic acid). • Each pack contains 6 cartridges.