Learn More
QA Bio Inc LudgerSep R BPT Solvent
Supplier:  QA Bio Inc LSRBPTX10
Encompass
The LudgerSep R BPTX Solvent was developed for the analysis of monosaccharides labeled with 2-AA using the LudgerTag Monosaccharide Labeling Kit • The LudgerSep R BPT Solvent is supplied at a 10x concentration of final as 50 mL of x10 BPT solvent in a rectangular HDPE bottle with a leak-proof cap. • The R BPT Solvent contains volatile components so we always recommend making the solvent up on the day of intended use.
Catalog No. 50-223-6723