Learn More
QA Bio Inc LudgerTag 2-AB, 2PB Reductant Glycan Labeling Kit
Supplier:  QA Bio Inc LTKABVP24
Encompass
For labelling of free glycans with 2-aminobenzamide acid (2-AB) using 2-picoline borane reductant. • Kit sufficient for 24 samples.
Catalog No. 50-223-6731