Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ludox™ Am Colloidal Silica

Catalog No. 42087518L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
4 L
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-087-518L 18 L
42-087-54L 4 L
42-087-51L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 42-087-518L Supplier Sigma Aldrich Supplier No. 42087518L
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

contains sodium stabilizing counterionLUDOX™ AM colloidal silica can be used as a source of silica for the synthesis of:NaX zeolites by hydrothermal method.Chrysotile-type Mg3Si2O5-+(OH)4 nanotubes (MgSi-NTs).It can also be used as a binder for ceramic slurries.LUDOX is a registered trademark of W.R. Grace and Co.-Conn.

Specifications

CAS 7631-86-9
Density 1.21 g/mL at 25°C
Molecular Formula O2Si
Linear Formula SiO2
Quantity 18 L
MDL Number MFCD00011232
Synonym Silica preparation
Molecular Weight (g/mol) 60.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.