Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ludox™ Cl Colloidal Silica

Catalog No. 4208831L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-088-31L 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 42-088-31L Supplier Sigma Aldrich Supplier No. 4208831L
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Alumina coated chloride stabilizing counterionLUDOX is a registered trademark of W.R. Grace and Co.-Conn.

Specifications

CAS 7440-43-9
Melting Point 320.9°C
Density 8.65 g/mL at 25°C
Boiling Point 765°C
Molecular Formula Cd
Linear Formula Cd
Quantity 1L
MDL Number MFCD00010914
Synonym Silica preparation
Molecular Weight (g/mol) 112.41 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.