Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Elbow, Male Luer to Female Luer Adapter; 25/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4550884
Encompass
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
Catalog No. NC0831384