Learn More
IDEX Health & Science Upchurch LUER ADAPTER FEMALE TO 10-32 M
Catalog No. NC1284277
Encompass_Preferred
Luer Adapter female luer to 10-32 male, peek
Catalog No. NC1284277 Supplier IDEX Health & Science Upchurch Supplier No. P719
Luer Adapter female luer to 10-32 male, peek