Learn More
Med Vet International Nipro Luer Adapter (Multi-Sample), 100/Box
Catalog No. 502092432
Encompass_Preferred
Designed for optimum efficiency: Luer Adapter allows for multiple draws with a single venipuncture. Accommodates needles sizes ranging from 23G to 19G to meet a wide spectrum of patient needs Dual capability permits use for either blood collection or a venous injection. Proprietary back end needle design facilitates smooth penetration of rubber tube stopper.
Catalog No. 50-209-2432 Supplier Med Vet International Supplier No. LM20G
Designed for optimum efficiency: Luer Adapter allows for multiple draws with a single venipuncture. Accommodates needles sizes ranging from 23G to 19G to meet a wide spectrum of patient needs Dual capability permits use for either blood collection or a venous injection. Proprietary back end needle design facilitates smooth penetration of rubber tube stopper.