Learn More
Trajan Scientific VICI Valco Luer Adapter, Female, to 1/4-28 Male, 1.50mm Bore, PEEK, Makes Leak-tight Connection from Male or Female Luer to 1/4-28 Threads
Catalog No. 501090210
Encompass
VICI Valco Luer Adapter, Female, to 1/4-28 Male, 1.50mm Bore, PEEK, Makes Leak-tight Connection from Male or Female Luer to 1/4-28 Threads. Made in the USA.
Catalog No. 50-109-0210 Supplier Trajan Scientific Supplier No. CFLAPK
VICI Valco Luer Adapter, Female, to 1/4-28 Male, 1.50mm Bore, PEEK, Makes Leak-tight Connection from Male or Female Luer to 1/4-28 Threads. Made in the USA.