Learn More
Ibidi USA Luer Connector Male, 25 pieces per box
SDP
Supplier:  Ibidi USA 10824
Encompass
Luer Connector Male connects Female Luers with 0.5, 0.8, or 1.6 mm ID tubing. polypropylene, sterile, 25 pcs./box
Catalog No. NC0994595