Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Elbow, Male Luer x Female Luer; 25/PK
Catalog No. NC0347553
Encompass
Easily complete your luer fittings systems with these premium connectors. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available.
Catalog No. NC0347553 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4550284
Easily complete your luer fittings systems with these premium connectors. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available.