Learn More
IDEX Health & Science Upchurch LUER FEM-MALE 1/4-28 TEFZEL
Catalog No. NC1919691
Encompass_Preferred
Idex Quick Connect Adapter, Red, Male Luer Adapter, For Use With: Pair to 1/4-28 Female, Temperature Max: 40 deg.C, Diameter Inner: 0.05 in., ETFE
Catalog No. NC1919691 Supplier IDEX Health & Science Upchurch Supplier No. P675
Idex Quick Connect Adapter, Red, Male Luer Adapter, For Use With: Pair to 1/4-28 Female, Temperature Max: 40 deg.C, Diameter Inner: 0.05 in., ETFE