Learn More
IDEX Health & Science Upchurch LUER FEM-MALE M6 TEFZEL
Catalog No. NC1919701
Encompass_Preferred
Idex Quick Connect Adapter, Black, Male Luer Adapter, For Use With: Pair to Female M6x1, Temperature Max: 80 deg.C, Diameter Inner: 0.05 in., ETFE
Catalog No. NC1919701 Supplier IDEX Health & Science Upchurch Supplier No. P677
Idex Quick Connect Adapter, Black, Male Luer Adapter, For Use With: Pair to Female M6x1, Temperature Max: 80 deg.C, Diameter Inner: 0.05 in., ETFE