Learn More
IDEX Health & Science Upchurch LUER FEMALE
Catalog No. NC1919068
Encompass_Preferred
Idex Adapters for HPLC, Connections: Female internal threads Luer to 1/4-28 Flat-bottom, For Use With: Female (Internal) Threads Luer to 1/4-28 Flat-Bottom, Male(External) Threads, Delrin.
Catalog No. NC1919068 Supplier IDEX Health & Science Upchurch Supplier No. P604
Idex Adapters for HPLC, Connections: Female internal threads Luer to 1/4-28 Flat-bottom, For Use With: Female (Internal) Threads Luer to 1/4-28 Flat-Bottom, Male(External) Threads, Delrin.