Learn More
IDEX Health & Science Upchurch LUER FEMALE PP 1/8IN THRUHOLE
Catalog No. NC1919461
Encompass_Preferred
Idex Threaded Luer Polypropylene Adapter, Diameter Inner: 0.125 mm, For Use With: Female to Male, Natural, Luer Adapter, Temperature Max: 40 deg.C
Catalog No. NC1919461 Supplier IDEX Health & Science Upchurch Supplier No. P618L
Idex Threaded Luer Polypropylene Adapter, Diameter Inner: 0.125 mm, For Use With: Female to Male, Natural, Luer Adapter, Temperature Max: 40 deg.C