Learn More
IDEX Health & Science Upchurch LUER FEMALE/UNION FEMALE 1/8IN
Catalog No. NC1919086
Encompass_Preferred
Idex LuerTight Fittings: Tefzel, Diameter Inner: 0.04 in., Natural, For Use With: To Female 1/4-28, Luer Adapter, ETFE, Temperature Max: 80 deg.C
Catalog No. NC1919086 Supplier IDEX Health & Science Upchurch Supplier No. P628
Idex LuerTight Fittings: Tefzel, Diameter Inner: 0.04 in., Natural, For Use With: To Female 1/4-28, Luer Adapter, ETFE, Temperature Max: 80 deg.C