Learn More
IDEX Health & Science Upchurch LUER FEMALE/UNON FEMALE 1/16IN
Catalog No. NC1919088
Encompass_Preferred
Idex LuerTight Fittings: Tefzel, Diameter Inner: 0.04 in., Natural, For Use With: To Female 10-32, Luer Adapter, ETFE, Temperature Max: 80 deg.C
Catalog No. NC1919088 Supplier IDEX Health & Science Upchurch Supplier No. P629
Idex LuerTight Fittings: Tefzel, Diameter Inner: 0.04 in., Natural, For Use With: To Female 10-32, Luer Adapter, ETFE, Temperature Max: 80 deg.C