Learn More
AVANTOR MASTERFLEX (VWR LLC) Cadence Luer Fitting Adapter, nickel-plated brass, male luer lock to 1/4"" barb; 1/ea
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4150766
Encompass
Exceptional durability. Easy to disassemble for cleaning. Autoclavable.
Catalog No. NC0542714