Learn More
Chemglass Life Sciences Inner Luer Joint, on 6.5mm OD x 1.0mm ID Capillary
SDP
Catalog No. CG11201
Encompass_Preferred
Inner Luer Joint, on 6.5mm OD x 1.0mm ID Capillary. Inner joint manufactured to the same specifications found on medical and research syringes. Complies with Federal Specification CG-S-921b and USAST Standard Z70.1. Available tooled on three tubing sizes with the inner diameter being 1mm through the tip of the joint on all three sizes.
Catalog No. CG11201 Supplier Chemglass Life Sciences Supplier No. CG11201
Inner Luer Joint, on 6.5mm OD x 1.0mm ID Capillary. Inner joint manufactured to the same specifications found on medical and research syringes. Complies with Federal Specification CG-S-921b and USAST Standard Z70.1. Available tooled on three tubing sizes with the inner diameter being 1mm through the tip of the joint on all three sizes.