Learn More
Chemglass Life Sciences Outer Luer Joint, 6.5mm OD x 1.0mm ID Capillary
SDP
Catalog No. CG11301
Encompass_Preferred
Outer Luer Joint, 6.5mm OD x 1.0mm ID Capillary. Outer joint manufactured to the same specifications found on medical and research syringes. Complies with Federal Specification CG-S-921b and USAST Standard Z70.1. Available tooled on three tubing sizes.
Catalog No. CG11301 Supplier Chemglass Life Sciences Supplier No. CG11301
Outer Luer Joint, 6.5mm OD x 1.0mm ID Capillary. Outer joint manufactured to the same specifications found on medical and research syringes. Complies with Federal Specification CG-S-921b and USAST Standard Z70.1. Available tooled on three tubing sizes.