Learn More
Chemglass Life Sciences Outer Luer Joint, 8mm OD x 5.0mm ID Capillary
SDP
Catalog No. CG11303
Encompass_Preferred
Outer Luer Joint, 8mm OD x 5.0mm ID Capillary. Outer joint manufactured to the same specifications found on medical and research syringes. Complies with Federal Specification CG-S-921b and USAST Standard Z70.1. Available tooled on three tubing sizes.
Catalog No. CG11303 Supplier Chemglass Life Sciences Supplier No. CG11303
Outer Luer Joint, 8mm OD x 5.0mm ID Capillary. Outer joint manufactured to the same specifications found on medical and research syringes. Complies with Federal Specification CG-S-921b and USAST Standard Z70.1. Available tooled on three tubing sizes.