Learn More
Air-Tite Luer Lock BIMED 2-PART SYRINGE
Catalog No. 14817258 Shop All Air-Tite Products
Change view
Click to view available options
Connection Type:
Centric Luer Lock
Catalog No. Connection Type
14-817-258 Centric Luer Lock
1 options
Catalog No. 14-817-258 Supplier Air-Tite Supplier No. AL30BMD
Get 1 Extra Syringe When You Buy 3
View all promotions
Add to Cart
Add to Cart
Ergonomic, durable syringe designed for precision, free from pyrogens, PVC, and toxins
  • 30 ml Luer Lock BIMED 2-PART SYRINGE - Designed with ergonomics and durability in mind, this syringe provides smooth handling and precise fluid administration.
  • Its secure Luer Lock tip ensures tight connections and prevents leakage, making it reliable for a wide range of laboratory and veterinary applications only.
  • With clear, easy-to-read graduation markings, it delivers accurate measurement every time. Manufactured from high-quality medical-grade plastic, it is free from pyrogens, PVC, and toxins, ensuring maximum safety for sensitive procedures.
  • Available in both sterile and non-sterile versions, the 30 ml BIMED syringe is trusted in pharmaceutical, diagnostic, and veterinary laboratory environments where precision and sterility are essential.
Specifications
Material Polyethylene (Plunger), Polypropylene (Barrel)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Luer Lock Syringe
Disposable Reusable
Volume (Metric) 30 mL
For Use With (Equipment) Syringe Filter
Connection Type Centric Luer Lock
Graduated Yes
Sterility Sterile