Learn More
International Medical Industries (IMI) Luer Lock Cap (Box of 100)
Catalog No. NC1780840
Encompass
Prep-Seal™ Caps offer convenient, economical, high-quality solutions to providing closure to medication delivery devices such as Luer lock syringes. Luer Lock Syringe Caps comes in a convenient sterile tray of 10. If your compounding needs call for volume capping of syringes, the Prep-Seal Luer Lock Cap is the perfect solution. A unique “keyed” tray bottom lets you install caps quickly and efficiently with a simple twist of the syringe; further enhancing the aseptic technique. • Sterile • Non-Pyrogenic • 10 Units Per Tray •Box of 100 • Blue
This item is not returnable. View return policy
Catalog No. NC1780840 Supplier International Medical Industries (IMI) Supplier No. 57100B
Prep-Seal™ Caps offer convenient, economical, high-quality solutions to providing closure to medication delivery devices such as Luer lock syringes. Luer Lock Syringe Caps comes in a convenient sterile tray of 10. If your compounding needs call for volume capping of syringes, the Prep-Seal Luer Lock Cap is the perfect solution. A unique “keyed” tray bottom lets you install caps quickly and efficiently with a simple twist of the syringe; further enhancing the aseptic technique. • Sterile • Non-Pyrogenic • 10 Units Per Tray •Box of 100 • Blue