Learn More
Chemglass Life Sciences Luer Lock Connection, Sampler, 10mL Syringe, Bioreactor
SDP
Catalog No. NC2293226
Encompass
.The Sampler uses all Luer style connections, eliminating any needle sampling and risk of puncture injuries.No valves.The sampling valve is opened by the attachment of the sampling syringe and automatically closes once the syringe is removed. Use with any type of cell culture sampling such as spinner flasks, bioreactors or bags.Sampler is set up with a check valve that eliminates the chances of contamination.NOTE: The sampler is supplied assembled, but not sterilized.It must be autoclaved prior to use.Syringes are individually sterile strip packed.
This item is not returnable. View return policy
Catalog No. NC2293226 Supplier Chemglass Life Sciences Supplier No. CLS1399A01
.The Sampler uses all Luer style connections, eliminating any needle sampling and risk of puncture injuries.No valves.The sampling valve is opened by the attachment of the sampling syringe and automatically closes once the syringe is removed. Use with any type of cell culture sampling such as spinner flasks, bioreactors or bags.Sampler is set up with a check valve that eliminates the chances of contamination.NOTE: The sampler is supplied assembled, but not sterilized.It must be autoclaved prior to use.Syringes are individually sterile strip packed.