Learn More
Health Care Logistics Inc LUER LOCK SYRNG CAPS,RED,100/C
SDP
Catalog No. NC1814367
Encompass
Sterile Luer Lock Syringe CapsIdeal for volume capping of syringes during compounding. Sterile packaging tray provides immediate access to 10 sterile caps and allows users to open 2 caps at a time without compromising the remaining caps. This reduces the risk of contamination and provides an ideal solution to save time and money.Quantity - 10/tray, 10 trays/case, total of 100
This item is not returnable. View return policy
Catalog No. NC1814367 Supplier Health Care Logistics Inc Supplier No. 19159R
Sterile Luer Lock Syringe CapsIdeal for volume capping of syringes during compounding. Sterile packaging tray provides immediate access to 10 sterile caps and allows users to open 2 caps at a time without compromising the remaining caps. This reduces the risk of contamination and provides an ideal solution to save time and money.Quantity - 10/tray, 10 trays/case, total of 100