Learn More
AVANTOR MASTERFLEX (VWR LLC) Cadence Female luer lock x 1/4"" NPT, 316 SS, each - 41507-48
Catalog No. NC0687173
Encompass
Exceptional durability. Easy to disassemble for cleaning. Ideal for harsh chemical environments. Autoclavable.
Catalog No. NC0687173 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4150748
Exceptional durability. Easy to disassemble for cleaning. Ideal for harsh chemical environments. Autoclavable.