Learn More
Eldon James Luer-Lok Cap Male 100/Pk
SDP
Catalog No. NC0637781 Shop All Fisherbrand Products
Encompass_Preferred
This item has a minimum qty of 7 per supplier requirements.
Precision engineered for smooth positive sealing. Eldon James luers are Certified Laboratory Tested. Our luers have been tested for dimensional and performance requirements according to ISO 594-1 and 594-2 standards. This testing includes dimensional checks, as well as tests on the security of the assembled fittings for leakage under pressure and vacuum conditions
This item is not returnable. View return policy
Catalog No. NC0637781
This item has a minimum qty of 7 per supplier requirements.
Precision engineered for smooth positive sealing. Eldon James luers are Certified Laboratory Tested. Our luers have been tested for dimensional and performance requirements according to ISO 594-1 and 594-2 standards. This testing includes dimensional checks, as well as tests on the security of the assembled fittings for leakage under pressure and vacuum conditions