Learn More
Hach Company Luer-Lok Tip Syringe 3 cc
Supplier:  Hach Company 4321300
Encompass
Syringe-Luer-Lok Tip-individually packaged 3 ccDisposable syringes feature a Luer-Lok tip suited for connection with syringe filters or filter holders Marked graduations 3 cc individually packaged 1pk
Catalog No. 50-002-07323