Learn More
IDEX Health & Science Upchurch LUER MALE
Catalog No. NC1919069
Encompass_Preferred
Idex LuerTight Fittings: Acetal, White, Luer Adapter, Diameter Inner: 0.05 in., For Use With: To Male 1/4-28, Temperature Max: 60 deg.C.
Catalog No. NC1919069 Supplier IDEX Health & Science Upchurch Supplier No. P605
Idex LuerTight Fittings: Acetal, White, Luer Adapter, Diameter Inner: 0.05 in., For Use With: To Male 1/4-28, Temperature Max: 60 deg.C.