Learn More
Ibidi USA Luer Plug Male 25/box, Natural Polypropylene, sterilized
SDP
Catalog No. NC1973663
Encompass
Closes female Luer adapters, such as those found on an ibidi ¿-Slide. Simply and tightly seals ibidi channel ¿-Slides with Luer adapters. Natural Polypropylene, sterilized
This item is not returnable. View return policy
Catalog No. NC1973663 Supplier Ibidi USA Supplier No. 10822
Closes female Luer adapters, such as those found on an ibidi ¿-Slide. Simply and tightly seals ibidi channel ¿-Slides with Luer adapters. Natural Polypropylene, sterilized