Learn More
IDEX Health & Science Upchurch LUER POLY FEMALE
Catalog No. NC1919072
Encompass_Preferred
Idex Threaded Luer Polypropylene Adapter, Diameter Inner: 0.05 in., For Use With: To Male 1/4-28, Natural, Luer Adapter, Temperature Max: 40 deg.C.
Catalog No. NC1919072 Supplier IDEX Health & Science Upchurch Supplier No. P618
Idex Threaded Luer Polypropylene Adapter, Diameter Inner: 0.05 in., For Use With: To Male 1/4-28, Natural, Luer Adapter, Temperature Max: 40 deg.C.