Learn More
Fine Science Tools Luer Rongeur, Stainless Steel, Curved, 15 cm
SDP
Catalog No. NC1692333
Encompass
Tip Shape: Curved/ Cup Size: 4mm/ Alloy / Material: Stainless Steel/ Length: 15cm/ Joint Action: Single
This item is not returnable. View return policy
Catalog No. NC1692333 Supplier Fine Science Tools Supplier No. 1600115
Tip Shape: Curved/ Cup Size: 4mm/ Alloy / Material: Stainless Steel/ Length: 15cm/ Joint Action: Single