Learn More
Air-Tite Luer Slip BIMED 2-PART SYRINGE
Catalog No. 14817257 Shop All Air-Tite Products
Change view
Click to view available options
Connection Type:
Eccentric Luer Slip
Catalog No. Connection Type
14-817-257 Eccentric Luer Slip
1 options
Catalog No. 14-817-257 Supplier Air-Tite Supplier No. A30BMD
Get 1 Extra Syringe When You Buy 3
View all promotions
Add to Cart
Add to Cart
Durable syringe designed for precision, available sterile/non-sterile with clear scaling for reliable use in laboratory
  • 30 ml Luer Slip BIMED 2-PART SYRINGE - Designed with ergonomics and durability in mind, this syringe provides smooth handling and precise fluid administration.
  • Its secure Luer Lock tip ensures tight connections and prevents leakage, making it reliable for a wide range of laboratory and veterinary applications only.
  • With clear, easy-to-read graduation markings, it delivers accurate measurement every time. Manufactured from high-quality medical-grade plastic, it is free from pyrogens, PVC, and toxins, ensuring maximum safety for sensitive procedures.
  • Available in both sterile and non-sterile versions, the 30 ml BIMED syringe is trusted in pharmaceutical, diagnostic, and veterinary laboratory environments where precision and sterility are essential.
Specifications
Material Polypropylene (Barrel), Polyethylene (Plunger)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Luer Slip
Disposable Reusable
Volume (Metric) 30 mL
For Use With (Equipment) Syringe Filter
Connection Type Eccentric Luer Slip
Graduated Yes
Sterility Sterile