Learn More
Air-Tite Luer Slip Syringe, Bulk
Catalog No. 14817280 Shop All Air-Tite Products
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-817-280 10 mL
14-817-277 1 mL
14-817-272 3 mL
14-817-275 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-817-280 Supplier Air-Tite Supplier No. MEZB10LS
Add to Cart
Add to Cart
For laboratory and veterinary use only
  • Individually sterile packed disposable
Specifications
Autoclavable Not Autoclavable
Material Polypropylene (Barrel), Polyethylene (Plunger), Polyisoprene Rubber (Tip)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Luer Slip Syringe
Disposable Single-use
Quantity 1050 Pk.
Volume (Metric) 10 mL
Connection Type Centric Luer Slip
Graduated Yes
Sterility Non-sterile
Show More Show Less
General Lab and Vet use only