Learn More
IDEX Health & Science Upchurch LUER TEFZEL FEMALE W/10-32
Catalog No. NC1919096
Encompass_Preferred
Idex Adapters for HPLC, Connections: Female Luer to Male 10-32 Adapter, For Use With: Female Luer to Male 10-32 Adapter, ETFE, Diameter Inner: 0.05 in.
Catalog No. NC1919096 Supplier IDEX Health & Science Upchurch Supplier No. P642
Idex Adapters for HPLC, Connections: Female Luer to Male 10-32 Adapter, For Use With: Female Luer to Male 10-32 Adapter, ETFE, Diameter Inner: 0.05 in.