Learn More
IDEX Health & Science Upchurch LUER TEFZEL MALE
Catalog No. NC1919085
Encompass_Preferred
Idex Adapters for HPLC, Connections: Male(External) threads Luer to 1/4-28 Flat-bottom, For Use With: Male(External) Threads Luer to 1/4-28 Flat-Bottom, Male(External) Threads, ETFE, Natural, Diameter Inner: 0.04 in.
Catalog No. NC1919085 Supplier IDEX Health & Science Upchurch Supplier No. P625
Idex Adapters for HPLC, Connections: Male(External) threads Luer to 1/4-28 Flat-bottom, For Use With: Male(External) Threads Luer to 1/4-28 Flat-Bottom, Male(External) Threads, ETFE, Natural, Diameter Inner: 0.04 in.